Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PIP4K2B (aa 299-324) Control Fragment Recombinant Protein

Catalog No. RP100405
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100405 Supplier Invitrogen™ Supplier No. RP100405
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. The encoded protein sequence does not show similarity to other kinases, but the protein does exhibit kinase activity. Additionally, the encoded protein interacts with p55 TNF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78356
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 8396
Name Human PIP4K2B (aa 299-324) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol-5-phosphate 4-kinase B; AI848124; c11; diphosphoinositide kinase 2-beta; LOW QUALITY PROTEIN: phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol 5-phosphate 4-kinase type II beta; Phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol-4-phosphate 5-kinase, type II, beta; phosphatidylinositol-5-phosphate 4-kinase type 2 beta; phosphatidylinositol-5-phosphate 4-kinase type-2 beta; phosphatidylinositol-5-phosphate 4-kinase, type II, beta; phosphatidylinositol-phosphate kinase IIgamma; PI(5)P 4-kinase type II beta; PI5P4KB; PI5P4Kbeta; PIP4K2B; PIP4KII-beta; Pip5k2b; PIP5KIIB; PIP5KIIbeta; PIP5P4KB; PIPKIIgamma; PTDINS(4)P-5-kinase; ptdIns(5)P-4-kinase isoform 2-beta
Common Name PIP4K2B
Gene Symbol PIP4K2B
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EERAEDEECENDGVGGNLLCSYGTPP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.