Learn More
Sino Biological Human PITPNA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10346MM0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PITPN (rh PITPN; Catalog#15366-H08H; NP_006215.1; Met1-Asp270). PITPN specific IgG was purified by Human PITPN affinity chromatography.
Catalog No. 50-237-4387