Learn More
BIOHIPPO Human PIVKA-II Microsample
SDP
Catalog No. 5000313953
Encompass
Human PIVKA-II Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIVKA-II in serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 13 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PIVKA-II Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PIVKA-II Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PIVKA-II biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13953 Supplier BIOHIPPO Supplier No. BHE15210680
Human PIVKA-II Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIVKA-II in serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 13 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PIVKA-II Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PIVKA-II Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PIVKA-II biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color