Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PIWIL1 Synthetic Peptide

Catalog No. PEP1165 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1165 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1165 Supplier Invitrogen™ Supplier No. PEP1165
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 18 amino acids near the amino terminus of human PIWI-L1. PEP-1165 can be used as a blocking peptide with polyclonal antibody PA5-21051.

Hiwi is a member of the Piwi /Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34- hematopoietic cells, suggesting a role for this gene in the maintenance of hematopoietic stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9271
Species Human
Purification Method purified
Accession Number Q96J94
Common Name PIWIL1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1
Gene Symbol PIWIL1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.