Learn More
BIOHIPPO Human PL Microsample
SDP
Catalog No. 5000314997
Encompass
Human PL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PL in serum plasma tissue homogenates and other biological fluids It provides a detection range of 2 5-160 ng/mL with a sensitivity of 0 9 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14997 Supplier BIOHIPPO Supplier No. BHE15211437
Human PL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PL in serum plasma tissue homogenates and other biological fluids It provides a detection range of 2 5-160 ng/mL with a sensitivity of 0 9 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color