Learn More
BIOHIPPO Human PLA2G12B Microsample
SDP
Catalog No. 5000314057
Encompass
Human PLA2G12B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PLA2G12B in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PLA2G12B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PLA2G12B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PLA2G12B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14057 Supplier BIOHIPPO Supplier No. BHE15211197
Human PLA2G12B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PLA2G12B in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PLA2G12B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PLA2G12B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PLA2G12B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color