Learn More
Proteintech Group Inc Human PLA2G1B ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0009896T
Encompass_Preferred
Phospholipase A2 group IB, also know as PLA2G1B, is a secreted member of the phospholipase A2 (PLA2) class of enzymes, is produced by the pancreatic acinar cells. PLA2G1B is a calcium-dependent enzyme which can catalyzes the hydrolysis of the sn-2 position of membrane glycerophospholipids to release arachidonic acid (AA) and lysophospholipids. PLA2G1B is involved in several physiological processes including cell contraction, cell proliferation and pathological response.
Catalog No. 50-197-4060