Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PLA2G5 Full-length ORF (NP_000920.1, 1 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89957184 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-184 10 ug
89-957-185 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-957-184 Supplier Abnova™ Supplier No. H00005322P01S
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the secretory phospholipase A2 family. It is located in a tightly-linked cluster of secretory phospholipase A2 genes on chromosome 1. The encoded enzyme catalyzes the hydrolysis of membrane phospholipids to generate lysophospholipids and free fatty acids including arachidonic acid. It preferentially hydrolyzes linoleoyl-containing phosphatidylcholine substrates. Secretion of this enzyme is thought to induce inflammatory responses in neighboring cells. Alternatively spliced transcript variants have been found, but their full-length nature has not been determined. [provided by RefSeq]

Sequence: MKGLLPLAWFLACSVPAVQGGLLDLKSMIEKVTGKNALTNYGFYGCYCGWGGRGTPKDGTDWCCWAHDHCYGRLEEKGCNIRTQSYKYRFAWGVVTCEPGPFCHVNLCACDRKLVYCLKRNLRSYNPQYQYFPNILCS

Specifications

Accession Number NP_000920.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5322
Molecular Weight (g/mol) 42.1kDa
Name PLA2G5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MKGLLPLAWFLACSVPAVQGGLLDLKSMIEKVTGKNALTNYGFYGCYCGWGGRGTPKDGTDWCCWAHDHCYGRLEEKGCNIRTQSYKYRFAWGVVTCEPGPFCHVNLCACDRKLVYCLKRNLRSYNPQYQYFPNILCS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686C2294/GV-PLA2/MGC46205/PLA2-10/hVPLA(2)
Common Name PLA2G5
Gene Symbol PLA2G5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.