Learn More
Bioworld HUMAN PLACENTA GROWTH FACTO
Catalog No. 50253544
Encompass_Preferred
HUMAN PLACENTA GROWTH FACTOR-2 (RECOMBINANT): Recombinant Human Placenta Growth Factor-2 Source: Insect Cells Background: Placenta growth factor (PlGF), a homodimeric glycoprotein that is homologous to vascular endothelial growth fac...
Catalog No. 50-253-544 Supplier Bioworld Supplier No. 220604471
HUMAN PLACENTA GROWTH FACTOR-2 (RECOMBINANT): Recombinant Human Placenta Growth Factor-2 Source: Insect Cells Background: Placenta growth factor (PlGF), a homodimeric glycoprotein that is homologous to vascular endothelial growth fac...