Learn More
Biomatik Corporation Human Placenta Specific Protein 9 (PLAC9) ELISA Kit
Supplier:  Biomatik Corporation EKU06648
Encompass_Preferred
Human Placenta Specific Protein 9 (PLAC9) ELISA Kit for detecting PLAC9 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5JTB6. Gene ID: 219348. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5708