Learn More
Biomatik Corporation Human Placental Thrombin Inhibitor (PTI) ELISA Kit
Supplier:  Biomatik Corporation EKU06651
Encompass_Preferred
Human Placental Thrombin Inhibitor (PTI) ELISA Kit for detecting PTI in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35237. Gene ID: 5269. Target Synonyms: SERPINB6; CAP; PI6; SPI3; Serpin Peptidase Inhibitor Clade B Member 6; Cytoplasmic antiproteinase; Peptidase inhibitor 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8818