Learn More
Creative Biomart Ecallantide
Supplier:  Creative Biomart THP0015
Encompass_Preferred
The product is a recombinant 60-amino-acid protein produced in Pichia pastoris yeast cells that contains three intramolecular disulfide bonds. It shares sequence similarities with the naturally occurring human protein tissue-factor pathway inhibitor (TFPI), which is also known lipoprotein-associated coagulation inhibitor (LACI). The amino acid sequence of two compounds differ by seven amino acids.
Catalog No. 50-208-4066