Learn More
Haematologic Technologies Inc HUMAN PLASMIN 500UG
Catalog No. 50883340
Encompass
Coagulation protein, Human Plasmin, 500 micrograms, Native enzyme, Mode of action: Enzyme involved in fibrinolysis, Research use only, 50% (v/v) glycerol/water, store at -20 Celsius.
Catalog No. 50-883-340 Supplier Haematologic Technologies Inc Supplier No. HCPM0140
Coagulation protein, Human Plasmin, 500 micrograms, Native enzyme, Mode of action: Enzyme involved in fibrinolysis, Research use only, 50% (v/v) glycerol/water, store at -20 Celsius.