Learn More
Biomatik Corporation Human Plasminogen Activator Inhibitor 1 (PAI1) CLIA Kit
Supplier:  Biomatik Corporation EKU08876
Encompass_Preferred
Human Plasminogen Activator Inhibitor 1 (PAI1) CLIA Kit for detecting PAI1 in plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.96pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05121. Gene ID: 5054. Target Synonyms: SERPINE1; PLANH1; Serpin Peptidase Inhibitor Clade E Member 1; Nexin,Plasminogen Activator Inhibitor Type 1 Serpin E1; Endothelial plasminogen activator inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6965