Learn More
Biomatik Corporation Human Plasminogen Activator, Urokinase (uPA) CLIA Kit
Supplier:  Biomatik Corporation EKU08847
Encompass_Preferred
Human Plasminogen Activator, Urokinase (uPA) CLIA Kit for detecting uPA in plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.24pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P00749. Gene ID: 5328. Target Synonyms: PLAU; ATF; URK; UK; UP-A; Abbokinase; Urokinase-Type Plasminogen Activator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7435