Learn More
Biomatik Corporation Human Plastin 1 (PLS1) ELISA Kit
Supplier:  Biomatik Corporation EKU06678
Encompass_Preferred
Human Plastin 1 (PLS1) ELISA Kit for detecting PLS1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14651. Gene ID: 5357. Target Synonyms: I-plastin; Fimbrin; Intestine-specific plastin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6037