Learn More
Biomatik Corporation Human Platelet Activating Factor Acetylhydrolase 2 (PAFAH2) ELISA Kit
Supplier:  Biomatik Corporation EKU06681
Encompass_Preferred
Human Platelet Activating Factor Acetylhydrolase 2 (PAFAH2) ELISA Kit for detecting PAFAH2 in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.105ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99487. Gene ID: 5051. Target Synonyms: SD-PLA2; Serine-dependent phospholipase A2; Platelet-activating factor acetylhydrolase 2, cytoplasmic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8816