Learn More
American Research Products Inc Human platelet actiVating fact
Catalog No. 5000326527
Encompass
Human platelet actiVating factor PAF ELISA Kit from CUSABIO Detection Range 4 69 ng/mL-300 ng/mL Sensitivity 1 17 ng/mL Sample serum urine cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26527 Supplier American Research Products Inc Supplier No. CSBE07929H
Human platelet actiVating factor PAF ELISA Kit from CUSABIO Detection Range 4 69 ng/mL-300 ng/mL Sensitivity 1 17 ng/mL Sample serum urine cell culture supernates tissue homogenates cell lysates Method Sandwich