Learn More
Biomatik Corporation Human Platelet Activating Factor Receptor (PAFR) ELISA Kit
Supplier:  Biomatik Corporation EKU08419
Encompass_Preferred
Human Platelet Activating Factor Receptor (PAFR) ELISA Kit for detecting PAFR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25105. Gene ID: 5724. Target Synonyms: PTAFR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4854