Learn More
Biomatik Corporation Human Platelet Derived Growth Factor AB (PDGFAB) CLIA Kit
Supplier:  Biomatik Corporation EKU08870
Encompass_Preferred
Human Platelet Derived Growth Factor AB (PDGFAB) CLIA Kit for detecting PDGFAB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04085. Gene ID: 5154. Target Synonyms: PDGF-AB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8609