Learn More
Biomatik Corporation Human Platelet Derived Growth Factor D (PDGFD) CLIA Kit
Supplier:  Biomatik Corporation EKU09538
Encompass_Preferred
Human Platelet Derived Growth Factor D (PDGFD) CLIA Kit for detecting PDGFD in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 54.9-40,000pg/ml. Sensitivity: 25.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9GZP0. Gene ID: 80310. Target Synonyms: PDGF-D; IEGF; SCDGF-B; Spinal Cord Derived Growth Factor B; Iris-expressed growth factor; Platelet-derived growth factor D, receptor-binding form/latent form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4812