Learn More
Biomatik Corporation Human Platelet Factor 4 (PF4) CLIA Kit
Supplier:  Biomatik Corporation EKU08855
Encompass_Preferred
Human Platelet Factor 4 (PF4) CLIA Kit for detecting PF4 in serum, platelet-poor plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.41-300pg/ml. Sensitivity: 0.17pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02776. Gene ID: 5196. Target Synonyms: CXCL4; SCYB4; Chemokine C-X-C-Motif Ligand 4; Oncostatin-A; Iroplact. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6226