Learn More
STEMCELL Technologies Human Platelet Lysate, Fibrinogen-Depleted, Size: 50 mL
SDP
Catalog No. 501974377
Encompass
Human platelet lysate is a growth factor-rich cell culture supplement derived from healthy donor human platelets at U.S. Food and Drug Administration (FDA)-licensed blood centers. Multiple donor units are pooled during manufacturing to minimize lot-to-lot variability. Pharmaceutical-grade heparin derived from porcine intestine is used in the manufacturing process. The concentration of heparin in the final product is ≤ 2 IU/mL.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. 50-197-4377 Supplier STEMCELL Technologies Supplier No. 06963
Human platelet lysate is a growth factor-rich cell culture supplement derived from healthy donor human platelets at U.S. Food and Drug Administration (FDA)-licensed blood centers. Multiple donor units are pooled during manufacturing to minimize lot-to-lot variability. Pharmaceutical-grade heparin derived from porcine intestine is used in the manufacturing process. The concentration of heparin in the final product is ≤ 2 IU/mL.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.