Learn More
STEMCELL Technologies Human Platelet Lysate, Size: 100 mL
SDP
Catalog No. NC1526832
Encompass
Human platelet lysate is a growth factor-rich cell culture supplement derived from healthy donor human platelets at U.S. Food and Drug Administration (FDA)-licensed and Health Canada-licensed blood centers. Multiple donor units are pooled during manufacturing to minimize lot-to-lot variability. Addition of an anticoagulant (e.g. Heparin Solution; Catalog #07980) is required to inhibit coagulation.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. NC1526832 Supplier STEMCELL Technologies Supplier No. 06961
Human platelet lysate is a growth factor-rich cell culture supplement derived from healthy donor human platelets at U.S. Food and Drug Administration (FDA)-licensed and Health Canada-licensed blood centers. Multiple donor units are pooled during manufacturing to minimize lot-to-lot variability. Addition of an anticoagulant (e.g. Heparin Solution; Catalog #07980) is required to inhibit coagulation.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.