Learn More
Sino Biological Anti-Urokinase/uPA Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10815MM05H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Urokinase / PLAU (rh Urokinase / PLAU; Catalog#10815-H08H; NP_002649.1; Met1-Leu431). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1319