Learn More
Sino Biological Anti-Urokinase/uPA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10815MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human uPA / PLAU (rhPLAU; Catalog#10815-H08H; Met 1-Leu 431; NP_002649.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7651