Learn More
Sino Biological Anti-Urokinase/uPA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10815T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Urokinase / PLAU (rh Urokinase / PLAU; Catalog#10815-H08H; NP_002649.1; Met1-Leu431). Urokinase / PLAU specific IgG was purified by Human Urokinase / PLAU affinity chromatography.
Catalog No. 50-240-9020