Learn More
Sino Biological Human PLAUR MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10925MM0820
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PLAUR / CD87 / uPAR (rh PLAUR / CD87 / uPAR; Catalog#10925-H08H; Q03405-1; Met 1-Arg 303). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1826