Learn More
Sino Biological Anti-uPAR/PLAUR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10925RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PLAUR / CD87 (rh PLAUR; Catalog#10925-H08H; Q03405-1; Met1-Arg303). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-5067