Learn More
BIOHIPPO Human PLD Microsample
SDP
Catalog No. 5000314828
Encompass
Human PLD Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PLD in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 42 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PLD Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PLD Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PLD biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14828 Supplier BIOHIPPO Supplier No. BHE15211557
Human PLD Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PLD in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 42 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PLD Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PLD Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PLD biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color