Learn More
Raybiotech Inc Human Plexin B2 ELISA Kit
Supplier:  Raybiotech Inc ELHPLXB21
Encompass_Preferred
Sensitivity: 0.05 ng/ml.Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Human Plexin B2.
Catalog No. 50-194-9650