Learn More
ACROBiosystems ACROBiosystems Human Plexin B2 Protein, His Tag
Catalog No. 502107224
Encompass_Preferred
Recombinant Protein;100UG;Human Plexin B2, His Tag (PL2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 1160 (Accession # O15031-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 127.0 kDa. The protein migrates as 150-180 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Plexin B2, His Tag (Cat. No. PL2-H52H3) at 5 μg/mL (100 μL/well) can bind Human SEMA4D, Fc Tag (Cat. No. CD0-H5257) with a linear range of 0.039-1.25 μg/mL (QC tested).
Catalog No. 50-210-7224 Supplier ACROBiosystems Supplier No. PL2H52H3100UG
Recombinant Protein;100UG;Human Plexin B2, His Tag (PL2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 1160 (Accession # O15031-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 127.0 kDa. The protein migrates as 150-180 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Plexin B2, His Tag (Cat. No. PL2-H52H3) at 5 μg/mL (100 μL/well) can bind Human SEMA4D, Fc Tag (Cat. No. CD0-H5257) with a linear range of 0.039-1.25 μg/mL (QC tested).