Learn More
ACROBiosystems Human PLGF / PGF (19-149) Protein, GST Tag
Supplier:  ACROBiosystems PGFH52G3500UG
Encompass_Preferred
Recombinant Protein;500UG;Human PLGF (19-221), GST Tag (PGF-H52G3) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 149 (Accession # P49763-2).;This protein carries a GST tag at the C-terminus. The protein has a calculated MW of 43.5 kDa.
Catalog No. 50-205-4916