Learn More
ACROBiosystems Human PLGF / PGF (19-149) Protein, His Tag (MALS verified)
Catalog No. 502055084
Encompass_Preferred
Recombinant Protein;500UG;Human PLGF (19-149) Protein, His Tag (PGF-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 149 (Accession # P49763-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.6 kDa.
Catalog No. 50-205-5084 Supplier ACROBiosystems Supplier No. PGFH52H5500UG
Recombinant Protein;500UG;Human PLGF (19-149) Protein, His Tag (PGF-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 149 (Accession # P49763-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.6 kDa.