Learn More
ACROBiosystems Human PLGF / PGF (19-149) Protein, Fc Tag (MALS verified)
Catalog No. 502055061
Encompass_Preferred
Recombinant Protein;500UG;Human PLGF (19-149), Fc Tag (PGF-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 149 (Accession # P49763-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.2 kDa.
Catalog No. 50-205-5061 Supplier ACROBiosystems Supplier No. PGFH5256500UG
Recombinant Protein;500UG;Human PLGF (19-149), Fc Tag (PGF-H5256) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 149 (Accession # P49763-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.2 kDa.