Learn More
ACROBiosystems Human PLGF / PGF (19-221) Protein, Fc Tag (MALS verified)
Catalog No. 502055064
Encompass_Preferred
Recombinant Protein;20UG;Human PLGF (19-221), Fc Tag (PGF-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 221 (Accession # P49763-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF R1, His Tag (Cat. No. VE1-H5220) at 2 μg/mL (100 μL/well) can bind Human PLGF (19-221), Fc Tag (Cat. No. PGF-H5255) with a linear range of 0.3-5 ng/mL (QC tested).;Immobilized Human PLGF (19-221), Fc Tag (Cat. No. PGF-H5255) at 2 μg/mL (100 μL/well) can bind Human VEGF R1, His Tag (Cat. No. VE1-H5220) with a linear range of 2-20 ng/mL (Routinely tested).
Catalog No. 50-205-5064 Supplier ACROBiosystems Supplier No. PGFH525520UG
Recombinant Protein;20UG;Human PLGF (19-221), Fc Tag (PGF-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 221 (Accession # P49763-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF R1, His Tag (Cat. No. VE1-H5220) at 2 μg/mL (100 μL/well) can bind Human PLGF (19-221), Fc Tag (Cat. No. PGF-H5255) with a linear range of 0.3-5 ng/mL (QC tested).;Immobilized Human PLGF (19-221), Fc Tag (Cat. No. PGF-H5255) at 2 μg/mL (100 μL/well) can bind Human VEGF R1, His Tag (Cat. No. VE1-H5220) with a linear range of 2-20 ng/mL (Routinely tested).