Learn More
Sino Biological Recombinant Human PLTP Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11171H08HB20
Encompass_Preferred
A DNA sequence encoding the human PLTP isoform 1 (P55058-1) (Met1-Val493) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4808