Learn More
Sino Biological Anti-TEM7 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11736MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TEM7 / PLXDC1 (rh TEM7 / PLXDC1; Catalog#11736-H08H; AAH36059.1; Met1-Thr426). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4595