Learn More
Sino Biological Anti-PMVK Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14583RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PMVK / phosphomevalonate kinase (rh PMVK; Catalog#14583-H07E; Q15126; Met1-Leu192). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8675