Learn More
Sino Biological Human PNLIP MAb 100ul (Sino Biological)
Supplier:  Sino Biological 13564MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PNLIP (rh PNLIP; Catalog#13564-H08H; P16233; Met1-Cys465). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3276