Learn More
Sino Biological Human PNLIP Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786002
Encompass_Preferred
The Human PNLIP ELISA Pair Set is for the quantitative determination of Human PNLIP.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human PNLIP coated on a 96-well plate. Standards and samples are added to the wells, and any Human PNLIP present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human PNLIP polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human PNLIP present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86002 Supplier Sino Biological Supplier No. SEK1356415
The Human PNLIP ELISA Pair Set is for the quantitative determination of Human PNLIP.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human PNLIP coated on a 96-well plate. Standards and samples are added to the wells, and any Human PNLIP present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human PNLIP polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human PNLIP present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.