Learn More
Sino Biological Anti-PNLIP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13564RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PNLIP (rh PNLIP; Catalog#13564-H08H; P16233; Met1-Cys465). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8954