Learn More
Sino Biological Anti-PNLIPRP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13565RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PNLIPRP1 / PLRP1 (rh PNLIPRP1 / PLRP1; Catalog#13565-H08H; AAH25784.1; Met1-Cys467). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4212