Learn More
BIOHIPPO Human PNPLA2 Microsample
SDP
Catalog No. 5000314727
Encompass
Human PNPLA2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PNPLA2 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 137 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PNPLA2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PNPLA2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PNPLA2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14727 Supplier BIOHIPPO Supplier No. BHE15211272
Human PNPLA2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PNPLA2 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 137 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PNPLA2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PNPLA2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PNPLA2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color