Learn More
ACROBiosystems Human Podoplanin, His Tag (PON-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Leu 131 (Accession # Q86YL7-1).
Supplier:  ACROBiosystems PONH52H3100UG
Encompass_Preferred
Human Podoplanin, His Tag (PON-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Leu 131 (Accession # Q86YL7-1).
Catalog No. 50-225-4014