Learn More
Sino Biological Anti-PODXL Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105082T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PODXL, and purified by antigen affinity chromatography.
Catalog No. 50-240-8182