Learn More
Biomatik Corporation Human Poly A Binding Protein Cytoplasmic 1 Like Protein (PABPC1L) ELISA Kit
Supplier:  Biomatik Corporation EKU06732
Encompass_Preferred
Human Poly A Binding Protein Cytoplasmic 1 Like Protein (PABPC1L) ELISA Kit for detecting PABPC1L in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4VXU2. Target Synonyms: C20orf119; PABPC1L1; ePAB; Polyadenylate-Binding Protein 1-Like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5027