Learn More
Biomatik Corporation Human Poly A Specific Ribonuclease (PARN) ELISA Kit
Supplier:  Biomatik Corporation EKU06733
Encompass_Preferred
Human Poly A Specific Ribonuclease (PARN) ELISA Kit for detecting PARN in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95453. Gene ID: 5073. Target Synonyms: DAN; Deadenylation Nuclease; Deadenylating nuclease; Polyadenylate-specific ribonuclease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8814