Learn More
Biomatik Corporation Human Polyamine Modulated Factor 1 Binding Protein 1 (PMFBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU06736
Encompass_Preferred
Human Polyamine Modulated Factor 1 Binding Protein 1 (PMFBP1) ELISA Kit for detecting PMFBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8TBY8. Gene ID: 83449. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8813