Learn More
American Research Products Inc Human Polycomb compleX protein
Catalog No. 5000327529
Encompass
Human Polycomb compleX protein BMI-1 (BMI1) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27529 Supplier American Research Products Inc Supplier No. CSBEL002726HU
Human Polycomb compleX protein BMI-1 (BMI1) ELISA kit from CUSABIO Detection Range 25 pg/mL-1600 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich